Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270854.1 | NP_001257783.1 | 8 - 9 | 992 | 60 | |
NM_012672.3 | NP_036804.2 | 9 - 10 | 1100 | 60 | ||
XM_006251709.2 | XP_006251771.1 | 4 - 5 | 1013 | 60 | ||
XM_008770500.1 | XP_008768722.1 | 6 - 7 | 904 | 60 | ||
GenBank mRNA | DQ191165.1 | - | 6 - 7 | 847 | 60 | |
HM043807.1 | - | 5 - 6 | 889 | 60 | ||
J03819.1 | - | 8 - 9 | 988 | 60 | ||
J03933.1 | - | 8 - 9 | 989 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Rattus norvegicus Probe spans exons |