Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017034.1 | NP_058730.1 | 1 - 2 | 85 | 88 | |
GenBank mRNA | X63675.1 | - | 1 - 2 | 85 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |