Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017286.2 | NP_058982.1 | 7 - 8 | 1226 | 110 | |
GenBank mRNA | BC089100.1 | - | 7 - 8 | 1251 | 7307104 | 110 |
J05156.1 | - | 7 - 8 | 1246 | 110 | ||
M22615.1 | - | 2 - 3 | 197 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |