Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022703.3 | NP_073194.1 | 11 - 12 | 1007 | 150 | |
XM_006240996.1 | XP_006241058.1 | 11 - 12 | 1112 | 150 | ||
XM_006240997.1 | XP_006241059.1 | 11 - 12 | 1109 | 150 | ||
XM_006240998.2 | XP_006241060.1 | 11 - 12 | 2952 | 150 | ||
GenBank mRNA | AF368278.1 | - | 11 - 12 | 149 | ||
AJ222724.1 | - | 11 - 12 | 995 | 150 | ||
BC087642.1 | - | 11 - 12 | 1007 | 7308544 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |