Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130829.1 | NP_570842.1 | 61 | 90 | ||
GenBank mRNA | AB058889.1 | - | 61 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus |