Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138531.2 | NP_612540.1 | 1 - 2 | 47 | 72 | |
GenBank mRNA | BC061711.1 | - | 1 - 2 | 47 | 5597943 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |