Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199091.2 | NP_954522.1 | 5 - 6 | 322 | 83 | |
GenBank mRNA | BC063178.1 | - | 5 - 6 | 326 | 6890550 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |