Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023973.1 | NP_076463.1 | 7 - 8 | 669 | 122 | |
GenBank mRNA | AF312699.1 | - | 7 - 8 | 669 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |