Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034117.1 | NP_001029289.1 | 5 - 6 | 548 | 126 | |
NM_053739.2 | NP_446191.1 | 5 - 6 | 570 | 126 | ||
GenBank mRNA | AY033824.1 | - | 4 - 5 | 346 | 126 | |
BC074011.1 | - | 5 - 6 | 435 | 7100185 | 126 | |
CK600131.1 | - | 5 - 6 | 548 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |