Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172327.2 | NP_758830.2 | 4 - 5 | 652 | 77 | |
XM_006230699.2 | XP_006230761.1 | 5 - 6 | 850 | 77 | ||
XM_006230700.2 | XP_006230762.1 | 5 - 6 | 819 | 77 | ||
GenBank mRNA | AF543560.1 | - | 4 - 5 | 686 | 77 | |
BC089110.1 | - | 4 - 5 | 652 | 7311852 | 77 | |
FQ214270.1 | - | 4 - 5 | 760 | 77 | ||
FQ228146.1 | - | 4 - 5 | 680 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |