Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234308.2 | XP_006234370.1 | 5 - 6 | 1054 | 84 | |
XM_345363.7 | XP_345364.5 | 5 - 6 | 1048 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |