Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013160.1 | NP_001013178.1 | 3 - 4 | 630 | 88 | |
XM_006234856.2 | XP_006234918.2 | 3 - 4 | 846 | 88 | ||
GenBank mRNA | BC079281.1 | - | 4 - 5 | 807 | 88 | |
BC091171.1 | - | 3 - 4 | 630 | 7132390 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |