Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106492.1 | NP_001099962.1 | 1 - 2 | 163 | 71 | |
XM_006234638.2 | XP_006234700.1 | 1 - 2 | 212 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |