Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257136.2 | XP_006257198.1 | 5 - 6 | 461 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |