Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241037.2 | XP_006241099.2 | 8 - 9 | 1395 | 72 | |
XM_008765152.1 | XP_008763374.1 | 8 - 9 | 1413 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |