Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_342941.7 | XP_342942.5 | 8 - 9 | 1049 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |