Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106536.1 | NP_001100006.1 | 12 - 13 | 2032 | 93 | |
XM_006235525.2 | XP_006235587.1 | 12 - 13 | 2069 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Rattus norvegicus Probe spans exons |