Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106993.1 | NP_001100463.1 | 4 - 5 | 566 | 79 | |
XM_006246092.2 | XP_006246154.1 | 4 - 5 | 572 | 79 | ||
XM_006246093.2 | XP_006246155.1 | 4 - 5 | 563 | 79 | ||
XM_006246094.2 | XP_006246156.1 | 3 - 4 | 500 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |