Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013960.1 | NP_001013982.1 | 13 - 14 | 1173 | 81 | |
GenBank mRNA | BC085822.1 | - | 13 - 14 | 1173 | 7126662 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |