Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109017.1 | NP_001102487.1 | 3 - 4 | 526 | 121 | |
XM_006257030.2 | XP_006257092.1 | 3 - 4 | 249 | 121 | ||
XM_006257031.2 | XP_006257093.1 | 2 - 3 | 529 | 121 | ||
XM_006257032.2 | XP_006257094.1 | 3 - 4 | 596 | 121 | ||
XM_008773264.1 | XP_008771486.1 | 2 - 3 | 181 | 121 | ||
XM_008773265.1 | XP_008771487.1 | 3 - 4 | 630 | 121 | ||
XM_008773266.1 | XP_008771488.1 | 2 - 3 | 181 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |