Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107082.1 | NP_001100552.1 | 9 - 10 | 1109 | 87 | |
XM_008768798.1 | XP_008767020.1 | 9 - 10 | 1122 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |