Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012044.1 | NP_001012044.1 | 13 - 14 | 1539 | 70 | |
XM_006252318.2 | XP_006252380.1 | 13 - 14 | 1714 | 70 | ||
XM_006252319.2 | XP_006252381.1 | 15 - 16 | 1913 | 70 | ||
XM_006252320.2 | XP_006252382.1 | 15 - 16 | 1961 | 70 | ||
XM_006252322.2 | XP_006252384.1 | 13 - 14 | 3120 | 70 | ||
GenBank mRNA | BC083855.1 | - | 13 - 14 | 1539 | 7125327 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |