Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751518.3 | XP_003751566.1 | 7 - 8 | 1279 | 83 | |
XM_006252367.2 | XP_006252429.1 | 7 - 8 | 1317 | 83 | ||
XM_006252368.2 | XP_006252430.1 | 6 - 7 | 1169 | 83 | ||
XM_006252369.2 | XP_006252431.1 | 7 - 8 | 1279 | 83 | ||
XM_008770927.1 | XP_008769149.1 | 7 - 8 | 1008 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |