Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109208.1 | NP_001102678.1 | 8 - 9 | 837 | 75 | |
XM_006235352.2 | XP_006235414.1 | 7 - 8 | 739 | 75 | ||
GenBank mRNA | X60658.1 | - | 8 - 9 | 803 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |