Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198744.2 | NP_942039.1 | 6 - 7 | 670 | 75 | |
XM_006235754.2 | XP_006235816.1 | 7 - 8 | 696 | 75 | ||
XM_006235757.2 | XP_006235819.1 | 6 - 7 | 656 | 75 | ||
XM_006235760.2 | XP_006235822.1 | 5 - 6 | 624 | 75 | ||
XM_008762499.1 | XP_008760721.1 | 7 - 8 | 630 | 75 | ||
GenBank mRNA | BC059167.1 | - | 6 - 7 | 671 | 6890828 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |