Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nip30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014014.1 | NP_001014036.1 | 3 - 4 | 388 | 86 | |
XM_006255110.2 | XP_006255172.1 | 3 - 4 | 698 | 86 | ||
XM_006255111.2 | XP_006255173.1 | 3 - 4 | 465 | 86 | ||
XM_006255114.2 | XP_006255176.1 | 4 - 5 | 930 | 86 | ||
GenBank mRNA | BC079145.1 | - | 3 - 4 | 388 | 7128234 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |