Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759823.1 | XP_008758045.1 | 8 - 9 | 1322 | 63 | |
XM_008759824.1 | XP_008758046.1 | 7 - 8 | 1027 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |