Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005885.2 | NP_001005885.1 | 4 - 5 | 497 | 140 | |
XM_006229766.2 | XP_006229828.1 | 4 - 5 | 562 | 140 | ||
GenBank mRNA | AY325765.1 | - | 4 - 5 | 537 | 140 | |
AY380804.1 | - | 4 - 5 | 555 | 140 | ||
BC098002.1 | - | 4 - 5 | 497 | 7453525 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |