Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133564.1 | NP_598248.1 | 4 - 5 | 631 | 97 | |
XM_006235674.2 | XP_006235736.1 | 5 - 6 | 4474 | 97 | ||
GenBank mRNA | AJ302054.1 | - | 4 - 5 | 631 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |