Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037186.1 | NP_001032263.1 | 685 | 140 | ||
GenBank mRNA | BC100071.1 | - | 685 | 7322585 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |