Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232238.2 | XP_006232300.1 | 2 - 3 | 1186 | 102 | |
XM_006232240.2 | XP_006232302.1 | 4 - 5 | 915 | 102 | ||
XM_008760950.1 | XP_008759172.1 | 5 - 6 | 3103 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |