Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135011.1 | NP_001128483.1 | 2 - 3 | 305 | 99 | |
XM_008766876.1 | XP_008765098.1 | 2 - 3 | 869 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |