Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277272.1 | NP_001264201.1 | 529 | 65 | ||
XM_006229555.2 | XP_006229617.1 | 1018 | 65 | |||
GenBank mRNA | BC158688.1 | - | 461 | 8375819 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |