Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014008.1 | NP_001014030.1 | 581 | 145 | ||
XM_006253695.2 | XP_006253757.1 | 2451 | 145 | |||
GenBank mRNA | BC083711.1 | - | 581 | 7190620 | 145 |
Gene Symbol: NEWGENE_1549776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253707.2 | XP_006253769.1 | 4 - 5 | 2451 | 145 | |
XM_008771557.1 | XP_008769779.1 | 3 - 4 | 2527 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |