Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107647.2 | NP_001101117.1 | 2 - 3 | 268 | 78 | |
XM_006231836.2 | XP_006231898.1 | 2 - 3 | 696 | 78 | ||
XM_008760671.1 | XP_008758893.1 | 1 - 2 | 2891 | 78 | ||
XM_008760672.1 | XP_008758894.1 | 2 - 3 | 2743 | 78 | ||
XM_008760673.1 | XP_008758895.1 | 2 - 3 | 500 | 78 | ||
GenBank mRNA | BC131851.1 | - | 2 - 3 | 268 | 7189311 | 78 |
FQ211666.1 | - | 2 - 3 | 319 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |