Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034923.1 | NP_001030095.1 | 10 - 11 | 1584 | 76 | |
XM_006244314.2 | XP_006244376.1 | 10 - 11 | 1739 | 76 | ||
GenBank mRNA | BC105780.1 | - | 10 - 11 | 1584 | 7932299 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |