Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107518.1 | NP_001100988.1 | 6 - 7 | 928 | 104 | |
XM_006229292.2 | XP_006229354.1 | 8 - 9 | 1031 | 104 | ||
XM_006229295.2 | XP_006229357.1 | 6 - 7 | 742 | 104 | ||
XM_006229296.2 | XP_006229358.1 | 6 - 7 | 798 | 104 | ||
XM_006229297.2 | XP_006229359.1 | 5 - 6 | 787 | 104 | ||
XM_006229298.2 | XP_006229360.1 | 5 - 6 | 605 | 104 | ||
XM_006229299.2 | XP_006229361.1 | 5 - 6 | 677 | 104 | ||
XM_008759390.1 | XP_008757612.1 | 5 - 6 | 633 | 104 | ||
XM_008759391.1 | XP_008757613.1 | 5 - 6 | 609 | 104 | ||
XM_008759392.1 | XP_008757614.1 | 7 - 8 | 1192 | 104 | ||
GenBank mRNA | FQ222971.1 | - | 5 - 6 | 791 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |