Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234024.2 | XP_006234086.1 | 4 - 5 | 816 | 97 | |
GenBank mRNA | AF390073.1 | - | 1 - 2 | 68 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |