Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039034.1 | NP_001034123.1 | 11 - 12 | 1109 | 58 | |
XM_006233831.2 | XP_006233893.1 | 12 - 13 | 1249 | 58 | ||
GenBank mRNA | BC100156.1 | - | 11 - 12 | 1109 | 7462311 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |