Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107176.1 | NP_001100646.1 | 4 - 5 | 825 | 76 | |
XM_006249871.2 | XP_006249933.1 | 5 - 6 | 1672 | 76 | ||
XM_006249872.2 | XP_006249934.1 | 5 - 6 | 2231 | 76 | ||
XM_008769554.1 | XP_008767776.1 | 4 - 5 | 2058 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |