Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpa33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191829.1 | NP_001178758.1 | 4 - 5 | 689 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |