Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080887.2 | NP_543163.1 | 5 - 6 | 770 | 126 | |
XM_006254827.2 | XP_006254889.1 | 5 - 6 | 769 | 126 | ||
GenBank mRNA | AF140358.1 | - | 5 - 6 | 574 | 126 | |
BC098908.1 | - | 5 - 6 | 770 | 7386445 | 126 | |
FQ213590.1 | - | 5 - 6 | 741 | 126 | ||
FQ226669.1 | - | 5 - 6 | 719 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |