Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106327.1 | NP_001099797.1 | 2 - 3 | 480 | 96 | |
XM_006230750.2 | XP_006230812.1 | 2 - 3 | 556 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |