Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231152.2 | XP_006231214.2 | 13 - 14 | 8650 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Rattus norvegicus Probe spans exons |