Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256890.2 | XP_006256952.1 | 6 - 7 | 1045 | 87 | |
XM_008773168.1 | XP_008771390.1 | 6 - 7 | 1045 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |