Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245835.2 | XP_006245897.2 | 13 - 14 | 4847 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |