Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108973.1 | NP_001102443.1 | 9 - 10 | 1606 | 65 | |
GenBank mRNA | JF422064.1 | - | 9 - 10 | 1243 | 65 | |
JF422065.1 | - | 8 - 9 | 1192 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |