Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271383.1 | NP_001258312.1 | 2 - 3 | 359 | 83 | |
XM_008763851.1 | XP_008762073.1 | 2 - 3 | 379 | 83 | ||
XM_008763852.1 | XP_008762074.1 | 2 - 3 | 320 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |