Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107952.1 | NP_001101422.1 | 11 | 3618 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Amplicon spans exons and probe does not span exons |