Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107960.1 | NP_001101430.1 | 2 - 3 | 624 | 84 | |
XM_006238657.2 | XP_006238719.1 | 1 - 2 | 1540 | 84 | ||
XM_006238658.2 | XP_006238720.1 | 2 - 3 | 1045 | 84 | ||
XM_006238659.2 | XP_006238721.1 | 2 - 3 | 2542 | 84 | ||
XM_008763977.1 | XP_008762199.1 | 1 - 2 | 1540 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |